Dibutyltin oxide for synthesis, Sigma-Aldrich®
: Merck
:818-08-6
:
:C₈H₁₈OSn
:MFCD00001992
:248.94 g/mol
:> 300 °C
:1.6 g/cm³ (20 °C)
:212-449-1
:3077
:9,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
:Ambient
- Solubility:0.004 g/l (20 °C)
- Molar Mass:248.94 g/mol
- Bulk Density:650 - 750 kg/m3
- Boiling Point:300 °C decomposes
- Density:1.5 g/cm3 (20 °C)
- Ignition Point:525 °C
- Hazard Class:9
- Hazard Class Description:Miscellaneous hazardous material
Frequently Bought Together