
Silicon(IV) oxide, amorphous ≥99.9% (metals basis), nanopowder
Catalog # 44781.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:60.08 g/mol
Merck Index:14,8493
Boiling Pt:2230 °C
Melting Pt:1710 °C
Density:2.4
Storage Temperature:Ambient
EINECS:231-545-4
:GHS08
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232