0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Lead (II) nitrate
Lead(II) nitrate ≥99.0% ACS
Lead(II) nitrate ≥99.0% ACS
:
Thermo Fisher Scientific
Lead(II) nitrate ≥99.0% ACS
:
Thermo Fisher Scientific
:
10099-74-8
:
:
Pb(NO₃)₂
:
MFCD00011153
:
331.21 g/mol
:
14,5414
:
470 °C (dec.)
:
4.530
:
233-245-9
:
1469
:
5.1,II
:
GHS03
:
GHS08
:
GHS07
:
Ambient
msds_ALFA014243
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
VWR®, Nitrile Gloves
: VWR Collection
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Hydrochloric acid 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., NF, Reag. USP volumetric solution
: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals