Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate, EMSURE® for analysis, Supelco®
Magnesium sulphate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Magnesium sulphate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    300 g/l (20 °C)
  • Molar Mass:
    120.37 g/mol
  • Bulk Density:
    600 kg/m3
  • Density:
    2.66 g/cm3 (20 °C)
  • pH:
    7.9 (50 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together