Magnesium sulphate
Supplier: LOHMANN
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Frequently Bought Together