Order Entry
Belgium
ContactUsLinkComponent
Calcium hydrogen orthophosphate, anhydrous ≥98.5% (by complexometric analysis), Puriss. p.a. analytical reagent, Fluka™
Calcium hydrogen orthophosphate, anhydrous ≥98.5% (by complexometric analysis), Puriss. p.a. analytical reagent, Fluka™
Catalogus # HONC21177-25KG
Leverancier:  Honeywell Chemicals
CAS-nummer:  
Calcium hydrogen orthophosphate, anhydrous ≥98.5% (by complexometric analysis), Puriss. p.a. analytical reagent, Fluka™
Catalogus # HONC21177-25KG
Leverancier:  Honeywell Chemicals
CAS-nummer:  

Specificaties

  • Pack type:
    Plastic bottle
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specificaties

Specificatie testresultaten

  • Assay:
    98.5 - 101.0 %
  • Loss on ignition (850°C):
    6.5 - 8.5 %
  • Insoluble in hydrochloric acid:
    Max. 0.2 %
  • Arsenic (As):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 200 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 2000 ppm
  • Manganese (Mn):
    Max. 100 ppm
  • Sodium (Na):
    Max. 500 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 200 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 1000 ppm
  • Chloride (Cl):
    Max. 100 ppm

Specificatie testresultaten

Over dit artikel

Synoniemen: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic
 :
 :136.06 g/mol
Merck index:13,01697
Smelten Pt:>450 °C
Dichtheid:2,89 g/cm³ (20 °C)
Temperatuur bij opslag:Ambient
EINECS:231-826-1
CAS-nummer:7757-93-9
Formule:CaHPO₄
MDL-nummer:MFCD00010909