

Sodium metabisulphite
Supplier: Roth Carl
Certificates
About this item
CAS Number:7681-57-4
:
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
:190.11 g/mol
Merck Index:12,08784
Melting Pt:150 °C
Density:1.48 g/cm³ (20 °C)
EINECS:231-673-0
REACH:01-2119531326-45
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together