Potassium dihydrogen phosphate ≥99%, crystallised, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:~100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 50 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 0.02 %
- Fe (Iron):Max. 100 ppm
Specification Test Results
Frequently Bought Together