Order Entry
Belgium
ContactUsLinkComponent
Potassium dihydrogen phosphate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium dihydrogen phosphate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium dihydrogen phosphate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 99.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (KH₂PO₄) (by acidimetry):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.01 %
  • Loss on Drying at 105°C:
    ≤0.2 %
  • pH of 5% Solution at 25°C:
    4.1 - 4.5
  • Chloride (Cl):
    ≤0.001 %
  • Fluoride (F):
    ≤0.001 %
  • Nitrogen Compounds (as N):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Iron (Fe):
    ≤0.002 %
  • Lead (Pb):
    ≤0.001 %
  • Sodium (Na):
    ≤0.005 %
  • Trace Impurities - Arsenic (As):
    ≤3.000 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together