
Potassium dihydrogen phosphate ≥99.0%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (KH₂PO₄):≥99.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:4.1 - 4.5
- Insoluble Matter, Calcium and NH₄OH Precipitate:≤0.01 %
- Loss on Drying at 105°C:≤0.2 %
- Heavy Metals (as Pb):≤0.001 %
- Iron (Fe):≤0.002 %
- Trace Impurities - Arsenic (As):≤3.000 ppm
Specification Test Results
Frequently Bought Together