Order Entry
Belgium
ContactUsLinkComponent
di-Potassium hydrogen phosphate ≥98% ACS, Fluka™
di-Potassium hydrogen phosphate ≥98% ACS, Fluka™
Certificates
undefined
di-Potassium hydrogen phosphate ≥98% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Loss on drying (105°C):
    Max. 1.0 %
  • pH (5 %, 25 °C):
    8.5 - 9.6
  • Iron (Fe):
    Max. 10 ppm
  • Sodium (Na):
    Max. 500 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 30 ppm
  • Total N:
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Appearance of the solution:
    complying

Specification Test Results

Related Information

Frequently Bought Together