Order Entry
Belgium
ContactUsLinkComponent
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • pH (20°C; 5 %):
    8.7 - 9.3
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Loss on drying (130°C):
    Max. 1 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 30 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together