Order Entry
Belgium
ContactUsLinkComponent
di-Potassium hydrogen phosphate, EMSURE® for analysis, Supelco®
di-Potassium hydrogen phosphate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
di-Potassium hydrogen phosphate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1600 g/l (20 °C)
  • Molar Mass:
    174.18 g/mol
  • Bulk Density:
    700 - 1000 kg/m3
  • Density:
    2.44 g/cm3 (20 °C)
  • pH:
    9 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together