di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
: Avantor
:7758-11-4
:
:K₂HPO₄
:MFCD00011383
:174.18 g/mol
:13,07743
:340 °C
:2.3 g/cm³ (20 °C)
:Ambient
:231-834-5
- GMP Manufactured Product:98.0 - 100.5%
- Meets U.S.P Requirements:≤3 ppm
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Pharmaceutical Chemical:≤0.03%
- USP - Assay (K₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤3 ppm
- USP - Carbonate (CO₃):Passes Test
- USP - Chloride (Cl):≤0.03 %
- USP - Fluoride (F):≤0.001 %
- USP - Identification:Passes Test
- USP - Insoluble Substances:≤0.2 %
- USP - Iron (Fe):≤0.003 %
- USP - Loss on Drying:≤1.0 %
- USP - Mono- or Tri- Basic Salt:Passes Test
- USP - pH (1 in 20):8.5 - 9.6
- USP - Sodium (Na):Passes Test
- USP - Sulfate (SO₄):≤0.1 %
Frequently Bought Together