Order Entry
Belgium
ContactUsLinkComponent
 
 
Sodium hydroxide ≥97.0%, pellets ACS, Fluka™
 
 
Sodium hydroxide ≥97.0%, pellets ACS, Fluka™
 

 

  • Assay:
    Min. 97.0 %
  • Assay of Na₂CO₃:
    Max. 1.0 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.001 %
  • Mercury (Hg):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.02 %
  • Magnesium (Mg):
    Max. 0.002 %
  • Nickel (Ni):
    Max. 0.001 %
  • Heavy metals (as Ag):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.005 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.003 %
  • Total N:
    Max. 0.001 %
  • Vapor Pressure:
    <18 mmHg ( 20 °C)

 

 

Frequently Bought Together