Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Corrosive
Marketsource
Supplier:   Roth Carl
Sodium hydroxide
Supplier:  Roth Carl
Marketsource
Certificates
Sodium hydroxide

2 Options Available Below

Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 23,90 €
    23,90 €
    1 * 500 g
       23,90 €

    Specifications:
    Marketsource
    Marketsource
  • Plastic bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 40,50 €
    40,50 €
    1 * 1 kg
       40,50 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.