Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide, pellets, pure, Supelco®
Potassium hydroxide, pellets, pure, Supelco®
Supplier:  Merck
Certificates
Potassium hydroxide, pellets, pure, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    1130 g/l (20 °C)
  • Melting Point:
    360 °C
  • Molar Mass:
    56.11 g/mol
  • Boiling Point:
    1320 °C
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.04 g/cm3 (20 °C)
  • pH:
    14 (56 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together