Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide, Reagent Grade
Corrosive
Irritant
Potassium hydroxide, Reagent Grade
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Potassium hydroxide, Reagent Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Arsenic:
    ≤0.0003%
  • Heavy Metals:
    ≤0.003%
  • Moisture:
    10 – 17%
  • Potassium Carbonate:
    ≤3.5%
  • Purity:
    >85.0%

Specification Test Results

Related Information

Frequently Bought Together