Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Potassium nitrate

Oxidizing
Marketsource
Supplier:   Roth Carl
Potassium nitrate
Supplier:  Roth Carl
Marketsource
Certificates
Potassium nitrate

2 Options Available Below

Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 40,90 €
    40,90 €
    1 * 500 g
       40,90 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 79,90 €
    79,90 €
    1 * 1 kg
       79,90 €

    Specifications:
    Marketsource
    Marketsource