Order Entry
Belgium
ContactUsLinkComponent
Homogeniser, high-throughput, HOHTDG
Homogeniser, high-throughput, HOHTDG
Catalog # 431-0261
Supplier:  OHAUS
undefined
Homogeniser, high-throughput, HOHTDG
Catalog # 431-0261
Supplier:  OHAUS

Specifications

  • Capacity:
    1 deep well plate, 4 microplates or any sample fitting the 102×127×64 mm adjustable holder
  • Speed range:
    300 – 1600 min⁻¹
  • Description:
    High-throughput homogeniser, HOHTDG, digital, 230 V, 50/60 Hz, 3,1 A
  • Model:
    HOHTDG
  • Display:
    LED
  • Power rating:
    450 W
  • Timer:
    1 second – 10 minutes
  • Weight:
    34 kg
  • W×D×H:
    443×288×518 mm
  • Pk:
    1

Specifications

About this item

The HT (High Throughput) lysing bead mill laboratory homogeniser provides efficient solutions for grinding, lysing, pulverising, mixing and homogenising in sample preparation applications.

  • Made with a polyurethane elastomer top housing, cycoloy base and a lexan lid
  • Flexibility to handle multiple samples and tube configurations
  • Five pre-set programs optimised for speed and time
  • Increased efficiency with high-throughput sample processing
  • Easy-to-read LED display allows the operator to view speed and time settings
  • Microprocessor-controlled unit includes sample clamp and hex key

An easy-to-load tray securely locks the microplate or tube in place. The high speed linear motion (31 mm) processes samples quickly. Ideal for DNA, RNA and protein analysis (from mammalian tissue, plant, soil, seed, fungi, bacteria, virus and other cells), cell lysing, PCR and gene expression.