
Lithium iodide, anhydrous ≥98%
Catalog # STRE93-0382-25G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:133.85 g/mol
Merck Index:13,05557
Boiling Pt:1180 °C
Melting Pt:450 °C
Density:3.494
Storage Temperature:Ambient
EINECS:233-822-5
:GHS07
CAS Number:10377-51-2
Formula:ILi
MDL Number:MFCD00011092