
Sulphuric acid ≥95.0-95.5% Reag. Ph. Eur. for testing of high viscosity paraffins
Catalog # KRAF05701.3000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589