Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pesticide reference standard, Dibenzofuran 50 µg/ml in n-nonane, CIL
Pesticide reference standard, Dibenzofuran 50 µg/ml in n-nonane, CIL
PROOCILULM17121.2
:
LGC Standards PROMOCHEM
:
Pesticide reference standard, Dibenzofuran 50 µg/ml in n-nonane, CIL
PROOCILULM17121.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
132-64-9
Description:
Dibenzofuran 50 µg/ml in n-nonane
Formula:
C₁₂H₈O
Description:
Dibenzofuran