4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) ≥99% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:23978-09-8
:
Formula:C₁₈H₃₆N₂O₆
MDL Number:MFCD00005111
:376.49 g/mol
Melting Pt:69…71 °C
:M-Clarity™ quality level = MQ200
EINECS:245-962-4
:GHS05
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Purity (GC):≥ 99
- Identity (IR-spectrum):passes test
- Melting range (°C):70 - 73
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_810647
SDS
ProductPage_810647
Product specification
Frequently Bought Together