Order Entry
Belgium
ContactUsLinkComponent
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%
Irritant
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%
Certificates
undefined
Irritant
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 72 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together