Order Entry
Belgium
ContactUsLinkComponent
 
Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650455-1L
CAS Number:  
undefined
Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650455-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.02
  • Absorbance at 225 nm:
    Max. 0.17
  • Absorbance at 200 nm:
    Max. 1.00
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Explosion Limit:
    9 %
  • Autoignition Temperature:
    500 °F

Specification Test Results

About this item

 :
 :84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1.013 hPa)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:8.3 %(V)
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814