1,3-Cyclohexadiene
Glass bottle for solids, 25 g
Catalog # MOLE21827894-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Cyclohexa-1,3-diene
- CAS Number:592-57-4
- Formula:C₆H₈
- MDL Number:MFCD00001532
- Molecular Weight:80.13 g/mol
- Boiling Pt:80 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.84 g/cm³ (20 °C)
- Flash Pt:–8 °C
- Storage Temperature:Ambient
- EINECS:209-764-1
- UN:3295
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



