Order Entry
Belgium
ContactUsLinkComponent
2,5-Dichlorobiphenyl-[13C12] (PCB No. 9) 40 µg/ml in n-nonane (99% 13C), CIL
2,5-Dichlorobiphenyl-[13C12] (PCB No. 9) 40 µg/ml in n-nonane (99% 13C), CIL
Certificates
2,5-Dichlorobiphenyl-[13C12] (PCB No. 9) 40 µg/ml in n-nonane (99% 13C), CIL

Specifications

  • CAS No.:
    25569-80-6
  • Formula:
    C₁₂H₈Cl₂
  • Description:
    2,5-Dichlorobiphenyl-[13C12](PCB No. 9)

Specifications

Frequently Bought Together