Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol red

Phenol red

Marketsource
Supplier:   Serva
Phenol red
Supplier:  Serva
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:13,07329
  • Melting Pt:> 300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 19,00 €
    19,00 €
    1 * 5 g
       19,00 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource