Order Entry
Belgium
ContactUsLinkComponent
 
Phenolphthalein 98.5%, pure indicator grade
 
undefined
Phenolphthalein 98.5%, pure indicator grade

 

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    98.0 to 102.0 %
  • Loss on drying:
    ≤0.5 % (105°C)
  • Sulfated ash:
    ≤0.1 %
  • Acidity/alkalinity:
    (per Ph. Eur.) Passes test
  • Color of solution:
    (per Ph. Eur.) Passes test
  • Chloride (Cl):
    ≤100 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

 

 

Frequently Bought Together