Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Chlorophenol red

Glass bottle for solids, 5 g
Catalog # MOLEM88339115
Marketsource
Supplier:   Molekula
Chlorophenol red
Catalog # MOLEM88339115
Supplier:  Molekula
Marketsource
CAS Number:  4430-20-0
Chlorophenol red

Glass bottle for solids, 5 g

CAS Number:  4430-20-0
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • MDL Number:MFCD00005877
  • Molecular Weight:423.27 g/mol
  • Melting Pt:∼261 °C
  • Storage Temperature:Ambient
  • EINECS:224-619-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications