Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Phenylethanol ≥98%

2-Phenylethanol ≥98%

Toxic
Irritant
2-Phenylethanol ≥98%
Certificates
2-Phenylethanol ≥98%
2-Phenylethanol ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 60-12-8 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.021 Refractive Index: 1.5330 Appearance: Colourless Melting Point: -27° Boiling Point: 219° Beilstein Reference: 1905732 Merck Reference: 14,7224 UN No.: 2810 MDL No.: MFCD00002886

Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Merck Index:14,7224
  • Boiling Pt:219 °C
  • Melting Pt:–27 °C
  • Density:1.021
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:200-456-2
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.