Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxy-p-cresol
2-Methoxy-p-cresol ≥98%
2-Methoxy-p-cresol ≥98%
:
Thermo Fisher Scientific
2-Methoxy-p-cresol ≥98%
:
Thermo Fisher Scientific
:
93-51-6
:
:
C₈H₁₀O₂
:
MFCD00002378
:
138.17 g/mol
:
14,2571
:
220…222 °C
:
5…6 °C
:
1.092
:
99 °C (210 °F)
:
202-252-9
:
GHS07
:
Ambient
msds_ALFAA13603
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Ethanol, GPR RECTAPUR®
: VWR Chemicals
cis-Jasmone 95%
: Apollo Scientific