
2-Methoxy-p-cresol 99%
:93-51-6
:
:C₈H₁₀O₂
:MFCD00002378
:138.17 g/mol
:13,02599
:219…222 °C (1013 hPa)
:2 °C
:1.09 g/cm³
:90 °C
:202-252-9
:GHS07
:Ambient
- Appearance:Clear colorless to light yellow Liquid (May turn yellow on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5350 to 1.5400 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together