p-Tolyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:622-59-3
:
Formula:C₈H₇NS
MDL Number:MFCD00004813
:149.22 g/mol
Boiling Pt:236…238 °C
Melting Pt:23…26 °C
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
EINECS:210-745-5
UN:2923
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together