m-Tolyl isothiocyanate for synthesis, Sigma-Aldrich®
Catalog # 8.18739.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Molar Mass:149.22 g/mol
- Boiling Point:243 °C (1013 hPa)
- Flash Point:106 °C
- Density:1.10 g/cm3 (20 °C)
Specification Test Results
About this item
:
:149.22 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:–20 °C
Density:1.103 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-677-6
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
:GHS05
CAS Number:621-30-7
Formula:C₈H₇NS
MDL Number:MFCD00004807