o-Tolyl isothiocyanate for synthesis
Catalog # 8.18738.0005
Supplier: Merck
CAS Number:
Specifications
- Seq:2210
Specifications
Specification Test Results
- Molar Mass:149.22 g/mol
- Boiling Point:140 - 143 °C (27 hPa)
- Flash Point:108 °C
- Density:1.11 g/cm3 (20 °C)
Specification Test Results
About this item
:149.22 g/mol
:M-Clarity™ quality level = MQ200
:GHS05
Storage Temperature:Ambient
Formula:C₈H₇NS
MDL Number:MFCD00004802