o-Tolyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Formula:C₈H₇NS
MDL Number:MFCD00004802
:149.22 g/mol
Flash Pt:107 °C (224 °F)
:GHS06
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together