Order Entry
Belgium
ContactUsLinkComponent
 
p-Tolyl isocyanate 99%
 
undefined
p-Tolyl isocyanate 99%

 

  • Appearance:
    Clear light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5301 to 1.5321 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together