N,N-Dimethyl-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99-97-8
:
Formula:C₉H₁₃N
MDL Number:MFCD00008316
:135.21 g/mol
Boiling Pt:211°C
Melting Pt:–15 °C
Density:0,9 g/cm³ (20 °C)
Flash Pt:90°C
Storage Temperature:Ambient
EINECS:202-805-4
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together