Benzoyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:532-55-8
:
Formula:C₈H₅NOS
MDL Number:MFCD00004815
:163.2 g/mol
Merck Index:14,1114
Boiling Pt:128…131 °C (15 mmHg)
Melting Pt:–1 °C
Density:1.214
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:208-540-0
UN:2922
ADR:8,III
:GHS06
:GHS08
:GHS05
Related Information
Frequently Bought Together