Order Entry
Belgium
ContactUsLinkComponent
(2-Chlorophenyl)acetonitrile for synthesis
(2-Chlorophenyl)acetonitrile for synthesis
Catalog # 8.18643.0010
Supplier:  Merck
CAS Number:  
(2-Chlorophenyl)acetonitrile for synthesis
Catalog # 8.18643.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Boiling Pt:241 °C (1013 hPa)
Melting Pt:21…24 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,II
 :151.6 g/mol
EINECS:220-669-4
UN:3439
 :GHS07
CAS Number:2856-63-5
Formula:C₈H₆ClN
MDL Number:MFCD00001898