Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Hydroquinone

Corrosive
Health
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Hydroquinone
Supplier:  Roth Carl
Marketsource
Certificates
Hydroquinone

3 Options Available Below

Product Details & Documents
Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,04833
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:172 °C
  • Density:1.332 g/cm³ (20 °C)
  • Flash Pt:165 °C
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource