Thiomersal
Supplier: Serva
Certificates
About this item
Synonyms: Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt
CAS Number:54-64-8
:
Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
MDL Number:MFCD00013062
:404.82 g/mol
Merck Index:12,09451
Melting Pt:232…233 °C
Flash Pt:250 °C (closed cup)
EINECS:200-210-4
UN:2025
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together