

Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:54-21-7
:
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440
:160.10 g/mol
Merck Index:12,08819
Melting Pt:200 °C
Density:0.32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:200-198-0
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Melting Point:200 °C
- Molar Mass:160.10 g/mol
- Bulk Density:320 kg/m3
- Density:0.32 g/cm3 (20 °C)
- pH:6.5 (100 g/l, H2O, 20 °C)
- Ignition Point:250 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06601.1000_TDS_EN ProductPage_106601
Product specification
msds_106601
SDS
Frequently Bought Together