Diethyl phthalate ≥98%, TECHNICAL
Metal drum, 25 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Phthalic acid diethyl ester
- CAS Number:84-66-2
- Formula:C₁₂H₁₄O₄
- MDL Number:MFCD00009111
- Molecular Weight:222.24 g/mol
- Merck Index:12,07527
- Boiling Pt:298 °C (1013 hPa)
- Melting Pt:–3 °C
- Density:1.12 g/cm³ (20 °C)
- Flash Pt:156 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:201-550-6
Documents
Certificates
Search by Lot Number30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
spec_VWRC23995.467 spec_VWRC23995.467 spec_VWRC23995.467
Product specification
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pack type:Metal drum
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



