Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Diethyl phthalate ≥99.5%, GPR RECTAPUR®

Plastic bottle, 5 L
Catalog # 23395.362
Supplier:   VWR Chemicals
Diethyl phthalate ≥99.5%, GPR RECTAPUR®
Catalog # 23395.362
Supplier:  VWR Chemicals
CAS Number:  84-66-2
Diethyl phthalate ≥99.5%, GPR RECTAPUR®
Diethyl phthalate ≥99.5%, GPR RECTAPUR®

Plastic bottle, 5 L

CAS Number:  84-66-2
Price
252,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Phthalic acid diethyl ester
  • CAS Number:84-66-2
  • Formula:C₁₂H₁₄O₄
  • MDL Number:MFCD00009111
  • Molecular Weight:222.24 g/mol
  • Merck Index:12,07527
  • Boiling Pt:298 °C (1013 hPa)
  • Melting Pt:–3 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:156 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:201-550-6
Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Density (20/4):
    1.110 - 1.120
  • Water:
    Max. 0.1 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications