Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methylparaben, Reference Standard

Methylparaben, Reference Standard

Methylparaben, USP reference standard, Vial, 125 mg
Catalog # 1432005.
Supplier:   USP
Methylparaben, Reference Standard
Catalog # 1432005.
Supplier:  USP
CAS Number:  
Methylparaben, Reference Standard

Methylparaben, USP reference standard, Vial, 125 mg

CAS Number:  
Price
366,00 €
Log in for availability
1 * 125 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    125 mg
  • Standards:
    USP reference standard
  • Description:
    Methylparaben
  • Cat. No.:
    1432005.
  • Formula:
    HOC₆H4CO2CH₃
  • Description:
    Methylparaben
  • Storage temperature:
    Ambient
  • Melting point:
    125…128 °C (lit)
  • CAS No.:
    99-76-3

Specifications