Supelco® Organic Reference Standard, Pentachlorophenol
Pentachlorophenol, Analytical standard, 1
Catalog # 48555-U
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338
Specifications
Specifications
- Pk:1
- Standards:Analytical standard
- Description:Pentachlorophenol
- Cat. No.:48555-U
- M-Clarity quality level:MQ100
- Formula:C₆Cl₅OH
- Storage conditions:2...30 °C
- Boiling point:310 °C (lit.)
- Hazard class description:H301 + H311 - H315 - H319 - H330 - H335 - H351 - H410
- MDL No.:MFCD00002162
- Melting point:165 - 180 °C (lit.)
- Density:1,978 g/ml at 25 °C (lit.)
- Molecular weight:266,34
- CAS No.:87-86-5
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



