2-Chlorobenzamide
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:609-66-5
:
Formula:C₇H₆ClNO
MDL Number:MFCD00007973
:155.58 g/mol
Melting Pt:141…145 °C
Density:1,34 g/cm³ (18 °C)
EINECS:210-195-6
Frequently Bought Together